4-Nitrobenzyl chloroformate, 95%
4-Nitrobenzyl chloroformate, 95%
4-Nitrobenzyl chloroformate, 95%
Thermo Scientific Chemicals

4-Nitrobenzyl chloroformate, 95%

CAS: 4457-32-3 | C8H6ClNO4 | 215.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 376040250
Price (USD)
168.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
168.00
Each
Chemical Identifiers
CAS4457-32-3
IUPAC Name(4-nitrophenyl)methyl carbonochloridate
Molecular FormulaC8H6ClNO4
InChI KeyMHSGOABISYIYKP-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(COC(Cl)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid and/or chunks or liquid after melting
Appearance (Color)Off-white to yellow to beige
Titration Argentometric>=94 % (ex Cl)
Infrared spectrumConforms
Melting point30°C to 36°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only