Thermo Scientific Chemicals

4-Fluorophenylacetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 376120250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Fluorophenylacetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 376120250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 376120250
Price (USD)
Price: 523.00
Online price: 500.65
Your price:
Quantity
-

Chemical Identifiers

CAS
459-04-1
IUPAC Name
2-(4-fluorophenyl)acetyl chloride
Molecular Formula
C8H6ClFO
InChI Key
SIOJFYRPBYGHOO-UHFFFAOYSA-N
SMILES
FC1=CC=C(CC(Cl)=O)C=C1
Refractive index
1.5110 to 1.5130 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance (Color)
Clear yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text