3-bromopropionaldehyde dimethylacetal, 95%, Thermo Scientific Chemicals
3-bromopropionaldehyde dimethylacetal, 95%, Thermo Scientific Chemicals
3-bromopropionaldehyde dimethylacetal, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-bromopropionaldehyde dimethylacetal, 95%, Thermo Scientific Chemicals

CAS: 36255-44-4 | Molecular Formula: C5H11BrO2 | Molecular Weight: 183.045 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3762600505 gGlass bottle
Catalog number 376260050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36255-44-4
IUPAC Name3-bromo-1,1-dimethoxypropane
Molecular FormulaC5H11BrO2
InChI KeyODZZAIFAQLODKN-UHFFFAOYSA-N
SMILESCOC(CCBr)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=94 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4460 to 1.4500 (20°C, 589 nm)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only