Nerol, 97%
Nerol, 97%
Nerol, 97%
Thermo Scientific Chemicals

Nerol, 97%

CAS: 106-25-2 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
Packaging:
Glass bottle
Catalog number 376540250
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.50
Save 4.85
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
30.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.50
Save 4.85
Each
Chemical Identifiers
CAS106-25-2
IUPAC Name(2Z)-3,7-dimethylocta-2,6-dien-1-ol
Molecular FormulaC10H18O
InChI KeyGLZPCOQZEFWAFX-YFHOEESVSA-N
SMILESCC(C)=CCC\C(C)=C/CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4720 to 1.4790 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only