Quinaldine red, pure
Quinaldine red, pure
Quinaldine red, pure
Thermo Scientific Chemicals

Quinaldine red, pure

CAS: 117-92-0 | C21H23IN2 | 430.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number 377510050
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.30
Save 7.65
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.30
Save 7.65
Each
Chemical Identifiers
CAS117-92-0
IUPAC Name2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide
Molecular FormulaC21H23IN2
InChI KeyJOLANDVPGMEGLK-UHFFFAOYSA-M
SMILES[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to green-black to dark blue
Infrared spectrumConforms
Appearance (Form)Powder
UV spectrumComplies
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only