Pyrrole-3-carboxylic acid hydrate, 95%
Pyrrole-3-carboxylic acid hydrate, 95%
Pyrrole-3-carboxylic acid hydrate, 95%
Pyrrole-3-carboxylic acid hydrate, 95%
Thermo Scientific Chemicals

Pyrrole-3-carboxylic acid hydrate, 95%

CAS: 336100-46-0 | C5H5NO2 | 111.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3776500252.5 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 377650025
Price (USD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS336100-46-0
IUPAC Name1H-pyrrole-3-carboxylic acid
Molecular FormulaC5H5NO2
InChI KeyDOYOPBSXEIZLRE-UHFFFAOYSA-N
SMILESOC(=O)C1=CNC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Water=<14 %
Appearance (Color)Beige to brown
Titration with NaOH>=94 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only