2-Thiazolecarboxaldehyde, 98%
2-Thiazolecarboxaldehyde, 98%
2-Thiazolecarboxaldehyde, 98%
Thermo Scientific Chemicals

2-Thiazolecarboxaldehyde, 98%

CAS: 10200-59-6 | C4H3NOS | 113.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 377680010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10200-59-6
IUPAC Name1,3-thiazole-2-carbaldehyde
Molecular FormulaC4H3NOS
InChI KeyZGTFNNUASMWGTM-UHFFFAOYSA-N
SMILESO=CC1=NC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange-red to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5730 to 1.5760 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only