D(+)-Carvone, 98%, synthetic
D(+)-Carvone, 98%, synthetic
D(+)-Carvone, 98%, synthetic
Thermo Scientific Chemicals

D(+)-Carvone, 98%, synthetic

CAS: 2244-16-8 | C10H14O | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 377840250
Price (USD)
61.80
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
61.80
Each
Chemical Identifiers
CAS2244-16-8
IUPAC Name(5S)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
Molecular FormulaC10H14O
InChI KeyULDHMXUKGWMISQ-VIFPVBQESA-N
SMILESCC(=C)[C@H]1CC=C(C)C(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+52.0° to +62.0° (20°C, 589 nm) (neat)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only