Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™
Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™
Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™
Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™
Acros Organics

Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™

CAS: 7791-25-5 | Cl2O2S | 134.96 g/mol
Have Questions?
Catalog NumberQuantityPackaging
378158000
also known as 37815
800 mLAcroSeal™ Glass Bottle
Catalog number 378158000
also known as 37815
HSN 28273990
Price (INR)
46,798.00
Each
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (INR)
46,798.00
Each
Add to cart
Chemical Identifiers
CAS7791-25-5
IUPAC Namesulfuroyl dichloride
Molecular FormulaCl2O2S
InChI KeyYBBRCQOCSYXUOC-UHFFFAOYSA-N
SMILESClS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration Argentometric0.95 to 1.10 M
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only