1,3-bis(2,6-diisopropylphenyl)imidazolium chloride, 97%
1,3-bis(2,6-diisopropylphenyl)imidazolium chloride, 97%
1,3-bis(2,6-diisopropylphenyl)imidazolium chloride, 97%
Thermo Scientific Chemicals

1,3-bis(2,6-diisopropylphenyl)imidazolium chloride, 97%

CAS: 250285-32-6 | C27H37ClN2 | 425.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3783100202 gGlass bottle
Catalog number 378310020
Price (USD)
-
Quantity:
2 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS250285-32-6
IUPAC Name1,3-bis[2,6-bis(propan-2-yl)phenyl]-3H-1λ⁵-imidazol-1-ylium chloride
Molecular FormulaC27H37ClN2
InChI KeyAVJBQMXODCVJCJ-UHFFFAOYSA-M
SMILES[Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only