1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%
1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%
1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%
Thermo Scientific Chemicals

1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride, 90%

CAS: 258278-25-0 | C27H39ClN2 | 427.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 378340010
Price (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS258278-25-0
IUPAC Name1,3-bis[2,6-bis(propan-2-yl)phenyl]-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium chloride
Molecular FormulaC27H39ClN2
InChI KeyLWPXTYZKAWSRIP-UHFFFAOYSA-M
SMILES[Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only