2-Chloro-4-iodopyridine, 98%
2-Chloro-4-iodopyridine, 98%
2-Chloro-4-iodopyridine, 98%
Thermo Scientific Chemicals

2-Chloro-4-iodopyridine, 98%

CAS: 153034-86-7 | C5H3ClIN | 239.44 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3784200505 gGlass bottle
Catalog number 378420050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS153034-86-7
IUPAC Name2-chloro-4-iodopyridine
Molecular FormulaC5H3ClIN
InChI KeyKJKIPRQNFDUULB-UHFFFAOYSA-N
SMILESClC1=CC(I)=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to brown-yellow
GC>=97.5 %
Infrared spectrumConforms
Melting point41°C to 45°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only