Zinc chloride, 99.99%, (trace metal basis), anhydrous
Zinc chloride, 99.99%, (trace metal basis), anhydrous
Zinc chloride, 99.99%, (trace metal basis), anhydrous
Thermo Scientific Chemicals

Zinc chloride, 99.99%, (trace metal basis), anhydrous

CAS: 7646-85-7 | Cl2Zn | 136.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 380130050
Price (USD)
70.65
Online exclusive
78.10
Save 7.45
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
70.65
Online exclusive
78.10
Save 7.45
Each
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Total trace metal impurities=<0.01 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only