Disodium hydrogen phosphate heptahydrate, 98.0-100.5% (dried basis), USP, GMP, J.T.Baker™
Avantor™

Disodium hydrogen phosphate heptahydrate, 98.0-100.5% (dried basis), USP, GMP, J.T.Baker™

This product is GMP Manufactured and meets U.S.P Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
12 kg
Catalog number 3803-07
Price (USD)
-
Quantity:
12 kg

This product is GMP Manufactured and meets U.S.P Requirements.

Specifications
CAS7782-85-6
Chemical Name or MaterialDisodium Hydrogen Phosphate Heptahydrate
ColorWhite
Melting Point48°C
Molecular FormulaHNa2O4P·7H2O
PackagingPolyethylene pail, screw top
SynonymSodium phosphate dibasic heptahydrate, Sodium monohydrogen phosphate heptahydrate, Disodium hydrogenorthophosphate 7-hydrate, Disodium hydrogenorthophosphate heptahydrate, Sodium phosphate dibasic 7-hydrate
GradeUSP
Molecular Weight (g/mol)268.07
Percent Purity98.0-100.5%
Quantity12 kg
Unit SizeEach