Sodium phosphate dibasic, anhydrous, 98.0-100.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sodium phosphate dibasic, anhydrous, 98.0-100.5% (dried basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP manufactured and meets JPE, EP, and USP Specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
12 kg
Catalog number 3804-01
Price (USD)
-
Quantity:
500 g

This product is GMP manufactured and meets JPE, EP, and USP Specifications.

Specifications
CAS7558-79-4
Chemical Name or MaterialSodium phosphate dibasic
ColorWhite
Melting Point250°C
Molecular FormulaNa2HPO4
PackagingAmber glass bottle, wide mouth, tamper-evident cap
SynonymDisodium hydrogen phosphate, Disodium hydrogen orthophosphate
GradeMulti-Compendial
Molecular Weight (g/mol)141.96
Percent Purity98.0-100.5%
Quantity500 g
Unit SizeEach