Tetrakis(dimethylamino)zirconium
Tetrakis(dimethylamino)zirconium
Tetrakis(dimethylamino)zirconium
Thermo Scientific Chemicals

Tetrakis(dimethylamino)zirconium

CAS: 19756-04-8 | C8H24N4Zr | 267.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
View all
Glass bottle
Catalog number 380410010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19756-04-8
Molecular FormulaC8H24N4Zr
InChI KeyDWCMDRNGBIZOQL-UHFFFAOYSA-N
SMILES[Zr+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C
Molecular Weight (g/mol)267.53
SpecificationsSpecification SheetSpecification Sheet
NMR (1H-NMR)Consistent with structure
Appearance (Color)Yellow-green
Appearance (Form)Crystalline solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only