Anhydride 3-chlorophtalique, 95+ %, Thermo Scientific Chemicals
Anhydride 3-chlorophtalique, 95+ %, Thermo Scientific Chemicals
Anhydride 3-chlorophtalique, 95+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Anhydride 3-chlorophtalique, 95+ %, Thermo Scientific Chemicals

CAS: 117-21-5 | C8H3ClO3 | 182.56 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
1 g
5 g
25 g
Conditionnement:
Flacon en verre
Référence 381290250
Prix (EUR)
364,00
Each
Quantité:
25 g
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)
364,00
Each
Identifiants chimiques
CAS117-21-5
Appearance (Form)Crystalline powder or flakes and/or chunks
Appearance (Color)White to light yellow
Infrared spectrumConforms
Melting point122°C to 126°C
Titration by coupling>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only