1,1'-Ferrocenedimethanol, 98%, Thermo Scientific Chemicals
1,1'-Ferrocenedimethanol, 98%, Thermo Scientific Chemicals
1,1'-Ferrocenedimethanol, 98%, Thermo Scientific Chemicals
1,1'-Ferrocenedimethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1'-Ferrocenedimethanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 382250010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1291-48-1
Molecular FormulaC12H14FeO2
SMILES[Fe].OCc1cccc1.OCc1cccc1
Molecular Weight (g/mol)246.09
MDL NumberMFCD02091688
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)Yellow to orange to brown
Melting point104°C to 109°C
Infrared spectrumConforms
Elemental Analysis>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only