Triphenylphosphinecopper(I) hydride hexamer, 96%
Triphenylphosphinecopper(I) hydride hexamer, 96%
Triphenylphosphinecopper(I) hydride hexamer, 96%
Thermo Scientific Chemicals

Triphenylphosphinecopper(I) hydride hexamer, 96%

CAS: 33636-93-0 | C108H96Cu6P6 | 1961.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 382510010
Price (USD)
96.80
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
96.80
Each
Chemical Identifiers
CAS33636-93-0
IUPAC Namehexahydrogen [2,3,4,5,6-pentakis(triphenylphosphaniumyl)heptacyclo[2.2.0.0¹,³.0²,⁵.0²,⁶.0³,⁵.0⁴,⁶]hexacopper-1-yl]triphenylphosphanium
Molecular FormulaC108H96Cu6P6
InChI KeyIZZFKTUYPRVQID-UHFFFAOYSA-T
SMILES[H+].[H+].[H+].[H+].[H+].[H+].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)[Cu]123[Cu]45([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]11([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]22([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]34([P+](C3=CC=CC=C3)(C3=CC=CC=C3)C3=CC=CC=C3)[Cu]512[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to dark red
Appearance (Form)Powder or crystals
Elemental Analysis>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only