2-Amino-2-cyanoacetamide, 97+%
2-Amino-2-cyanoacetamide, 97+%
2-Amino-2-cyanoacetamide, 97+%
Thermo Scientific Chemicals

2-Amino-2-cyanoacetamide, 97+%

CAS: 6719-21-7 | C3H5N3O | 99.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 382760050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6719-21-7
IUPAC Name2-amino-2-cyanoacetamide
Molecular FormulaC3H5N3O
InChI KeyJRWAUKYINYWSTA-UHFFFAOYNA-N
SMILESNC(C#N)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to tan
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=97.0 %
Melting point120°C to 124°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only