Nalgene™ PETG Low Particulate Biotainer Bottles
Nalgene™ PETG Low Particulate Biotainer Bottles
Nalgene™ PETG Low Particulate Biotainer Bottles
Thermo Scientific™

Nalgene™ PETG Low Particulate Biotainer Bottles

Nalgene PETG Low Particulate Biotainer Bottles are certified clean to USP <788> standards, making them an excellent solution for biological and pharmaceutical manufacturing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)IncludesQuantity
383110-421 L, 1 L, 1 L, 1 LSide Handle, Side Handle, Side Handle, Side Handle7 x 5 Bottles, 7 x 5 Bottles, 7 x 5 Bottles, 7 x 5 Bottles
383025-42125 mL, 125 mL, 125 mL, 125 mLSide Handle, Side Handle, Side Handle, Side Handle20 x 5 Bottles, 20 x 5 Bottles, 20 x 5 Bottles, 20 x 5 Bottles
383005-42500 mL, 500 mL, 500 mL, 500 mLSide Handle, Side Handle, Side Handle, Side Handle14 x 5 Bottles, 14 x 5 Bottles, 14 x 5 Bottles, 14 x 5 Bottles
383230-422 L, 2 L, 2 L, 2 LSide Handle, Side Handle, Side Handle, Side Handle4 x 5 Bottles, 4 x 5 Bottles, 4 x 5 Bottles, 4 x 5 Bottles
383230-202 L, 2 L, 2 L, 2 LSide Handle, Side Handle, Side Handle, Side Handle20 Bottles, 20 Bottles, 20 Bottles, 20 Bottles
383415-165 L, 5 L, 5 L, 5 LSide Handle, Side Handle, Side Handle, Side Handle6 Bottles, 6 Bottles, 6 Bottles, 6 Bottles
383415-425 L, 5 L, 5 L, 5 LNeck Handle, Neck Handle, Neck Handle, Neck Handle6 Bottles, 6 Bottles, 6 Bottles, 6 Bottles
Catalog number 383110-42
Price (USD)
-
Capacity (Metric):
1 L, 1 L, 1 L, 1 L
Includes:
Side Handle, Side Handle, Side Handle, Side Handle
Quantity:
7 x 5 Bottles, 7 x 5 Bottles, 7 x 5 Bottles, 7 x 5 Bottles
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Certified Clean PETG Biotainer™ Bottles reduce the risk of carry-over contamination.
  • Specifically designed for pharmaceutical and biological manufacturing
  • Certified Clean, validated to USP <788>
  • <0.5 EU/mL Endotoxin
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AutoclavableNot Autoclavable
Capacity (English)33.814 fl. oz.
Certifications/ComplianceUSP <778>
Closure ColorWhite, White, White, White
Closure LinerLDPE, LDPE, LDPE, LDPE
Closure MaterialPolypropylene, Polypropylene, Polypropylene, Polypropylene
Closure Size48 mm, 48 mm, 48 mm, 48 mm
Diameter (Metric)98 mm
Diameter (Metric) Outer98 mm
Diameter (Metric) Outer Neck48 mm, 48 mm, 48 mm, 48 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Preparation ​, Media/Buffer Preparation ​, Media/Buffer Preparation ​, Media/Buffer Storage, Media/Buffer Storage, Media/Buffer Storage, Media/Buffer Storage, Sampling, Sampling, Sampling, Sampling, Storage, Storage, Storage, Storage, Transportation ​, Transportation ​, Transportation ​, Transportation ​, Validation Testing, Validation Testing, Validation Testing, Validation Testing, Drug Substance, Drug Substance, Drug Substance, Drug Substance, Drug Product, Drug Product, Drug Product, Drug Product
Height (Metric)201 mm, 201 mm, 201 mm, 201 mm
IncludesSide Handle, Side Handle, Side Handle, Side Handle
Length (English)27.75 in., 27.75 in., 27.75 in., 27.75 in.
Low ParticulateUSP <788> (Low particulate), USP <788> (Low particulate), USP <788> (Low particulate), USP <788> (Low particulate)
No. per Case35/Cs., 35/Cs., 35/Cs., 35/Cs.
No. per Pack5/Pk., 5/Pk., 5/Pk., 5/Pk.
Quantity7 x 5 Bottles, 7 x 5 Bottles, 7 x 5 Bottles, 7 x 5 Bottles
Width (English)20 in., 20 in., 20 in., 20 in.
Workflow StepUpstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
AutoclavableNot Autoclavable
Capacity (English)33.814 fl. oz.
Capacity (Metric)1 L, 1 L, 1 L, 1 L
Certifications/ComplianceUSP <778>
ColorClear, Clear, Clear, Clear
MaterialPET, PETG, PET, PETG
Product TypeSquare Bottle, Biotainer, Square Bottle, Biotainer
SterilitySterile, Sterile, Sterile, Sterile
Unit SizeEach