Samarium(III) trifluoromethanesulfonate, 98%
Samarium(III) trifluoromethanesulfonate, 98%
Samarium(III) trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Samarium(III) trifluoromethanesulfonate, 98%

CAS: 52093-28-4 | C3F9O9S3Sm | 597.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Plastic bottle
Catalog number 383170010
Price (USD)
-
Quantity:
1 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS52093-28-4
IUPAC Namesamarium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9O9S3Sm
InChI KeyDDCWGUIPLGMBPO-UHFFFAOYSA-K
SMILES[Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Elemental Analysis24.65 to 25.66 % (Sm)
Infrared spectrumConforms
Appearance (Form)Powder
Elemental Analysis5.91 to 6.15 % (C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only