Nitrosonium hexafluorophosphate, 95%
Nitrosonium hexafluorophosphate, 95%
Nitrosonium hexafluorophosphate, 95%
Thermo Scientific Chemicals

Nitrosonium hexafluorophosphate, 95%

CAS: 16921-91-8 | F6NOP | 174.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Plastic bottle
Catalog number 383730050
Price (USD)
139.00
Each
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
139.00
Each
Chemical Identifiers
CAS16921-91-8
IUPAC Namehexafluoro-λ⁵-phosphanuide; iminooxidanium
Molecular FormulaF6NOP
InChI KeySAKPNYRUBHJGAR-UHFFFAOYSA-N
SMILESN#[O+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Appearance (Color)Colorless to white
Assay>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only