Chloroform, 99+%, extra pure, stabilized with amylene
Chloroform, 99+%, extra pure, stabilized with amylene
Chloroform, 99+%, extra pure, stabilized with amylene
Thermo Scientific Chemicals

Chloroform, 99+%, extra pure, stabilized with amylene

CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog NumberQuantity
3837600505 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 383760050
Price (USD)
-
Quantity:
5 L
Request bulk or custom format
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
Color scale=<10 APHA
Residue after evaporation=<0.002 %
StabilizerApprox. 100 ppm Amylene
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only