Isohexane, for pesticide residue analysis, contains <5% n-Hexane
Isohexane, for pesticide residue analysis, contains <5% n-Hexane
Isohexane, for pesticide residue analysis, contains <5% n-Hexane
Thermo Scientific Chemicals

Isohexane, for pesticide residue analysis, contains <5% n-Hexane

CAS: 73513-42-5 | C6H14 | 86.18 g/mol
数量:
1 L
2.5 L
パッケージ:
Glass Bottle
製品番号(カタログ番号) 383820010
価格(JPY)
-
数量:
1 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS73513-42-5
IUPAC Name2-methylpentane
Molecular FormulaC6H14
InChI KeyAFABGHUZZDYHJO-UHFFFAOYSA-N
SMILESCCCC(C)C
さらに表示
Water=<0.01 % (Coulometric)
GC(ECD) Passes test
Appearance (Color)Clear colorless
Residue after evaporation=<2 ppm
GC(NPD) Passes test
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
RUO – Research Use Only