3,4-difluorobenzyl Chloride, 98%
3,4-difluorobenzyl Chloride, 98%
3,4-difluorobenzyl Chloride, 98%
Thermo Scientific Chemicals

3,4-difluorobenzyl Chloride, 98%

CAS: 698-80-6 | C7H5ClF2 | 162.56 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 384420050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS698-80-6
IUPAC Name4-(chloromethyl)-1,2-difluorobenzene
Molecular FormulaC7H5ClF2
InChI KeyVTGRVYJPIVZZGS-UHFFFAOYSA-N
SMILESFC1=CC=C(CCl)C=C1F
さらに表示
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.4920 to 1.4940 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only