1-(2-Hydroxyethyl)-4-methylpiperazine, 98%
1-(2-Hydroxyethyl)-4-methylpiperazine, 98%
1-(2-Hydroxyethyl)-4-methylpiperazine, 98%
Thermo Scientific Chemicals

1-(2-Hydroxyethyl)-4-methylpiperazine, 98%

CAS: 5464-12-0 | C7H16N2O | 144.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 384830050
Price (USD)
296.65
Special offer
Online exclusive
Ends: 28-Dec-2026
349.00
Save 52.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
296.65
Special offer
Online exclusive
Ends: 28-Dec-2026
349.00
Save 52.35
Each
Chemical Identifiers
CAS5464-12-0
IUPAC Name2-(4-methylpiperazin-1-yl)ethan-1-ol
Molecular FormulaC7H16N2O
InChI KeyQHTUMQYGZQYEOZ-UHFFFAOYSA-N
SMILESCN1CCN(CCO)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Low melting solid
Refractive index1.4850 to 1.4870 (20°C, 589 nm)
Appearance (Color)Light yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only