Propyl propionate, 98+%
Propyl propionate, 98+%
Propyl propionate, 98+%
Thermo Scientific Chemicals

Propyl propionate, 98+%

CAS: 106-36-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 385522500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-36-5
IUPAC Namepropyl propanoate
Molecular FormulaC6H12O2
InChI KeyMCSINKKTEDDPNK-UHFFFAOYSA-N
SMILESCCCOC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Refractive index1.3820 to 1.3940 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only