Glass Wool - FAQs
View additional product information for Glass Wool - FAQs (386060500, 386062500, 386060010)
1 product FAQs found
What is the diameter of Glass Wool (Cat. No. 386060010, 386060500, 386062500) threads?
Glass Wool (Cat. No. 386060010, 386060500, 386062500) threads have a diameter of 10-11 microns.