6-Bromo-2-pyridinecarboxaldehyde, 97%
6-Bromo-2-pyridinecarboxaldehyde, 97%
6-Bromo-2-pyridinecarboxaldehyde, 97%
Thermo Scientific Chemicals

6-Bromo-2-pyridinecarboxaldehyde, 97%

CAS: 34160-40-2 | C6H4BrNO | 186.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 387420050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS34160-40-2
IUPAC Name6-bromopyridine-2-carbaldehyde
Molecular FormulaC6H4BrNO
InChI KeyQWFHFNGMCPMOCD-UHFFFAOYSA-N
SMILESBrC1=CC=CC(C=O)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point81°C to 84°C
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)White to beige to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only