Bis(trifluoromethanesulfonyl)imide, 99%
Bis(trifluoromethanesulfonyl)imide, 99%
Bis(trifluoromethanesulfonyl)imide, 99%
Thermo Scientific Chemicals

Bis(trifluoromethanesulfonyl)imide, 99%

CAS: 82113-65-3 | C2HF6NO4S2 | 281.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 387600010
Price (EUR)
45,65
Online exclusive
50,50
Save 4,85 (10%)
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
45,65
Online exclusive
50,50
Save 4,85 (10%)
Each
Chemical Identifiers
CAS82113-65-3
IUPAC Name1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide
Molecular FormulaC2HF6NO4S2
InChI KeyZXMGHDIOOHOAAE-UHFFFAOYSA-N
SMILESFC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)Colorless to brown
Melting point50°C to 56°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only