Potassium phosphate, tribasic, 97%, pure, anhydrous
Potassium phosphate, tribasic, 97%, pure, anhydrous
Potassium phosphate, tribasic, 97%, pure, anhydrous
Thermo Scientific Chemicals

Potassium phosphate, tribasic, 97%, pure, anhydrous

CAS: 7778-53-2 | K3O4P | 212.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 387680100
Price (USD)
502.65
Online Exclusive
558.00
Save 55.35 (10%)
Each
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
502.65
Online Exclusive
558.00
Save 55.35 (10%)
Each
Chemical Identifiers
CAS7778-53-2
IUPAC Nametripotassium phosphate
Molecular FormulaK3O4P
InChI KeyLWIHDJKSTIGBAC-UHFFFAOYSA-K
SMILES[K+].[K+].[K+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess HCl>=96.0 %
Appearance (Color)White
Appearance (Form)Powder or granules and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only