Hydrochloric acid, pure, 25% solution in water
Hydrochloric acid, pure, 25% solution in water
Hydrochloric acid, pure, 25% solution in water
Thermo Scientific Chemicals

Hydrochloric acid, pure, 25% solution in water

CAS: 7647-01-0 | ClH | 36.46 g/mol
製品番号(カタログ番号) 387790010
価格(JPY)
-
数量:
1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
さらに表示
Appearance (Color)Clear colorless
Titration with NaOH>=25.0 %
Appearance (Form)Solution
Sulfate (SO4)=<0.002 %
Heavy metals (as Pb)=<1 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only