10-Undecen-1-ol, 98+%, stabilized
10-Undecen-1-ol, 98+%, stabilized
10-Undecen-1-ol, 98+%, stabilized
Thermo Scientific Chemicals

10-Undecen-1-ol, 98+%, stabilized

CAS: 112-43-6 | C11H22O | 170.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 388990250
Price (USD)
38.40
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.40
Each
Chemical Identifiers
CAS112-43-6
IUPAC Nameundec-10-en-1-ol
Molecular FormulaC11H22O
InChI KeyGIEMHYCMBGELGY-UHFFFAOYSA-N
SMILESOCCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4480 to 1.4530 (20°C, 589 nm)
StabilizerAbt 0.1% alpha-tocopherol (synthetic)
Appearance (Color)Clear colorless to light yellow
GC>=98.0 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only