Sodium cyclamate, 99%
Sodium cyclamate, 99%
Sodium cyclamate, 99%
Thermo Scientific Chemicals

Sodium cyclamate, 99%

CAS: 139-05-9 | C6H12NNaO3S | 201.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 389280010
Price (USD)
147.65
Special offer
Online exclusive
Ends: 28-Dec-2026
174.00
Save 26.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
147.65
Special offer
Online exclusive
Ends: 28-Dec-2026
174.00
Save 26.35
Each
Chemical Identifiers
CAS139-05-9
IUPAC Namesodium N-cyclohexylsulfamate
Molecular FormulaC6H12NNaO3S
InChI KeyUDIPTWFVPPPURJ-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO4>=98.5 % (On dry substance)
Loss on drying=<1 %
Sulfate (SO4)=<0.1 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only