Ammonium tetrathiomolybdate, 99.95%, (trace metal basis)
Ammonium tetrathiomolybdate, 99.95%, (trace metal basis)
Ammonium tetrathiomolybdate, 99.95%, (trace metal basis)
Thermo Scientific Chemicals

Ammonium tetrathiomolybdate, 99.95%, (trace metal basis)

CAS: 15060-55-6 | H10MoN2S4 | 262.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 389530010
Price (USD)
47.20
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.20
Each
Chemical Identifiers
CAS15060-55-6
IUPAC Namediammonium disulfanylidenemolybdenum disulfanide
Molecular FormulaH10MoN2S4
InChI KeyZKKLPDLKUGTPME-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Molybdenum (Mo)35.5 to 38.1 %
Total trace metal impurities=<500 ppm
Appearance (Color)Dark red or green or purple or black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only