Potassium polysulfide, pure
Potassium polysulfide, pure
Potassium polysulfide, pure
Thermo Scientific Chemicals

Potassium polysulfide, pure

CAS: 37199-66-9 | (S)nK2 | NaN g/mol
Have Questions?
Catalog NumberQuantityPackaging
3895900101 kgPlastic bottle
Catalog number 389590010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS37199-66-9
Molecular Formula(S)nK2
SMILES[K+].[K+].*-[S--]-*
MDL NumberMFCD00148074
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Ochre to brown
Appearance (Form)Lumps
Titration Iodometric42.0 to 44.0 % (K2S)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only