Rhodium(II) trifluoroacetate dimer
Rhodium(II) trifluoroacetate dimer
Rhodium(II) trifluoroacetate dimer
Thermo Scientific Chemicals

Rhodium(II) trifluoroacetate dimer

CAS: 31126-95-1 | C8F12O8Rh2 | 657.88 g/mol
数量:
250 mg
1 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 389702500
価格(JPY)
-
数量:
250 mg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS31126-95-1
IUPAC Nametetrakis(trifluoroacetic acid) dirhodium
Molecular FormulaC8H4F12O8Rh2
InChI KeySZQVMUPTZFMHQT-UHFFFAOYSA-N
SMILES[Rh].[Rh].OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F
さらに表示
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Rhodium (Rh)27.5 to 35.0 %
Appearance (Color)Green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only