Isopropanol, 99.5+%, extra pure
Isopropanol, 99.5+%, extra pure
Isopropanol, 99.5+%, extra pure
Thermo Scientific Chemicals

Isopropanol, 99.5+%, extra pure

CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 L
25 L
Packaging:
View all
Plastic Drum
Catalog number 389710100
Price (USD)
175.00
Each
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
175.00
Each
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Water=<0.2 %
Non-volatile matter=<0.001 %
GC>=99.50 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only