Acetonitrile-d3, for NMR, 99.5 atom % D
Thermo Scientific Chemicals

Acetonitrile-d3, for NMR, 99.5 atom % D

CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Catalog NumberQuantity
38986010010 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 389860100
Price (USD)
-
Quantity:
10 mL
Request bulk or custom format
Chemical Identifiers
CAS2206-26-0
IUPAC Name(²H₃)acetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES[2H]C([2H])([2H])C#N
View more
SpecificationsSpecification SheetSpecification Sheet
D-Enrichment>=99.50 %
Water=<0.05 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only