Iron(III) phosphate hydrate, extra pure
Iron(III) phosphate hydrate, extra pure
Iron(III) phosphate hydrate, extra pure
Thermo Scientific Chemicals

Iron(III) phosphate hydrate, extra pure

CAS: 51833-68-2 | FeH2O5P | 168.83 g/mol
Have Questions?
Catalog NumberQuantityPackaging
389955000500 gGlass bottle
Catalog number 389955000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS51833-68-2
IUPAC Nameiron(3+) hydrate phosphate
Molecular FormulaFeH2O5P
InChI KeyRQKNQWUJROKKEQ-UHFFFAOYSA-K
SMILESO.[Fe+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to pink to brown
Titration with Na2S2O326.0 to 32.0 % (as Fe)
Loss on ignition=<32.5 %
Arsenic (As)=<3 ppm
Fluoride (F)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only