Amberlite™ FPC3500, ion-exchange resin
Amberlite™ FPC3500, ion-exchange resin
Amberlite™ FPC3500, ion-exchange resin
Thermo Scientific Chemicals

Amberlite™ FPC3500, ion-exchange resin

CAS: 12125-02-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 389990010
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS950182-46-4
MDL NumberMFCD08460528
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Beads
Total volume capacity>=2.60 eq/l (H)
Particle size=<5.0 % (> 1.180 mm)
Harmonic mean size0.450 to 0.650 mm
Moisture holding capacity60 to 70 % (H)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only