Magnesium tert-butoxide, 93+%
Magnesium tert-butoxide, 93+%
Magnesium tert-butoxide, 93+%
Thermo Scientific Chemicals

Magnesium tert-butoxide, 93+%

CAS: 32149-57-8 | C8H18MgO2 | 170.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 390470500
Price (TWD)
-
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS32149-57-8
IUPAC Namemagnesium(2+) 2-methylpropan-2-olate
Molecular FormulaC4H9MgO
InChI KeyONFWJTUNDMIGEP-UHFFFAOYSA-N
SMILES[Mg++].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Titration with HCl>=93.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only