tert-Butyl methyl ether, for analysis
tert-Butyl methyl ether, for analysis
tert-Butyl methyl ether, for analysis
Thermo Scientific Chemicals

tert-Butyl methyl ether, for analysis

CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 390480010
Price (USD)
60.65
Special offer
Online exclusive
Ends: 28-Dec-2026
70.60
Save 9.95
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 28-Dec-2026
70.60
Save 9.95
Each
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.5 %
Peroxides=<5 ppm
Calcium (Ca)=<0.5 ppm
Copper (Cu)=<0.02 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only