1-Methyl-2-pyrrolidinone, 99+%, ACS reagent
1-Methyl-2-pyrrolidinone, 99+%, ACS reagent
1-Methyl-2-pyrrolidinone, 99+%, ACS reagent
1-Methyl-2-pyrrolidinone, 99+%, ACS reagent
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99+%, ACS reagent

CAS: 872-50-4 | C5H9NO | 99.133 g/mol
数量:
250 mL
2.5 L
10 L
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 390682500
価格(JPY)
-
数量:
250 mL
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
さらに表示
Water=<0.05 %
Nickel (Ni)=<0.02 ppm
Iron (Fe)=<0.1 ppm
Lead (Pb)=<0.1 ppm
Color scale=<50 APHA
さらに表示

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only