tert-Butanol, ACS reagent
tert-Butanol, ACS reagent
tert-Butanol, ACS reagent
Thermo Scientific Chemicals

tert-Butanol, ACS reagent

CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number 390690010
Price (USD)
114.00
Each
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
114.00
Each
Chemical Identifiers
CAS75-65-0
IUPAC Name2-methylpropan-2-ol
Molecular FormulaC4H10O
InChI KeyDKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILESCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Carbonyl compounds=<0.01 % (as formaldehyde)
Infrared spectrumConforms
Water=<0.1 % (K.F.)
Titrable acid=<0.001 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only