3-Butynyl p-toluenesulfonate, 96%
3-Butynyl p-toluenesulfonate, 96%
3-Butynyl p-toluenesulfonate, 96%
Thermo Scientific Chemicals

3-Butynyl p-toluenesulfonate, 96%

CAS: 23418-85-1 | C11H12O3S | 224.28 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 390780050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS23418-85-1
IUPAC Namebut-3-yn-1-yl 4-methylbenzene-1-sulfonate
Molecular FormulaC11H12O3S
InChI KeySTOASOOVVADOKH-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OCCC#C
さらに表示
Appearance (Form)Liquid
Refractive index1.5215 to 1.5255 (20°C, 589 nm)
Appearance (Color)Clear colorless to brown
Infrared spectrumConforms
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only