3-Butynyl p-toluenesulfonate, 96%
3-Butynyl p-toluenesulfonate, 96%
3-Butynyl p-toluenesulfonate, 96%
Thermo Scientific Chemicals

3-Butynyl p-toluenesulfonate, 96%

CAS: 23418-85-1 | C11H12O3S | 224.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 390780250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS23418-85-1
IUPAC Namebut-3-yn-1-yl 4-methylbenzene-1-sulfonate
Molecular FormulaC11H12O3S
InChI KeySTOASOOVVADOKH-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5215 to 1.5255 (20°C, 589 nm)
Appearance (Color)Clear colorless to brown
Infrared spectrumConforms
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only