Xylenes, extra pure, mixture of isomers
Xylenes, extra pure, mixture of isomers
Xylenes, extra pure, mixture of isomers
Xylenes, extra pure, mixture of isomers
Thermo Scientific Chemicals

Xylenes, extra pure, mixture of isomers

CAS: 1330-20-7 | C8H10
数量:
1 L
2.5 L
パッケージ:
Glass Bottle
製品番号(カタログ番号) 390790025
価格(JPY)
-
見積もりを依頼する
数量:
2.5 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
さらに表示
GC>=98.0 % (mixed isomers)
Substance darkened by H2SO4Passes test
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Water=<0.05 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only