3-Pyridinepropionic acid, 97%
3-Pyridinepropionic acid, 97%
3-Pyridinepropionic acid, 97%
Thermo Scientific Chemicals

3-Pyridinepropionic acid, 97%

CAS: 3724-19-4 | C8H9NO2 | 151.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
Catalog number 390980050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3724-19-4
IUPAC Name3-(pyridin-3-yl)propanoic acid
Molecular FormulaC8H9NO2
InChI KeyWDGXIUUWINKTGP-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and/or chunks
Melting point153°C to 157°C
Infrared spectrumConforms
Titration with NaOH>=96.0 %
Appearance (Color)White to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only